Gene Transcripts

Gene Symbol
MGEA5
Entrez Gene ID
10724
Gene Name
meningioma expressed antigen 5 (hyaluronidase)
Gene Aliases
MEA5, NCOAT, OGA
Chromosome Location
Chr. 10: 101784443 - 101818709 on Build GRCh38
UniGene ID
Hs.500842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142434.1 NP_001135906.1 15 4576
NM_012215.3 NP_036347.1 16 4735
XM_017015585.1 XP_016871074.1 17 5112
XM_017015586.1 XP_016871075.1 13 3899
GenBank mRNA AB014579.2 - 16 4297
AF036144.2 - 16 4687
AK027008.1 - 4 2092

View Full Product Details