Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252129.1 | NP_001239058.1 | 4 | 866 | |
NM_012321.4 | NP_036453.1 | 5 | 908 | ||
GenBank mRNA | AF117235.1 | - | 5 | 871 | |
AF182290.1 | - | 5 | 785 | ||
AF251218.1 | - | 5 | 779 | ||
AJ238096.1 | - | 5 | 760 | ||
BC000387.2 | - | 5 | 788 | 2821171 | |
BC003652.2 | - | 5 | 786 | 3536946 | |
BC022198.2 | - | 5 | 786 | 3534432 | |
BC023665.2 | - | 5 | 781 | 5015049 | |
BF432099.1 | - | 1 | 401 | ||
BU539495.1 | - | 1 | 328 | ||
CF147062.1 | - | 1 | 88 |