Gene Transcripts

Gene Symbol
Hnf1a
Entrez Gene ID
24817
Gene Name
HNF1 homeobox A
Gene Aliases
HNF1, LF-B1, Lfb1, Tcf1
Chromosome Location
Chr. 12: 47407811 - 47433342 on Build Rnor_6.0
UniGene ID
Rn.9660
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012669.1 NP_036801.1 4 984
GenBank mRNA J03170.1 - 4 984
X53297.1 - 3 342
X54423.1 - 4 834

View Full Product Details