Gene Transcripts

Gene Symbol
PRND
Entrez Gene ID
23627
Gene Name
prion protein 2 (dublet)
Gene Aliases
DOPPEL, DPL, PrPLP, dJ1068H6.4
Chromosome Location
Chr. 20: 4721854 - 4728462 on Build GRCh38
UniGene ID
Hs.406696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012409.3 NP_036541.2 2 423
GenBank mRNA AF187843.1 - 2 357
AF187844.1 - 1 308
AK313404.1 - 2 424
AY358985.1 - 2 352
BC043644.2 - 2 367 5272576

View Full Product Details