Gene Transcripts
- Gene Symbol
- Adrbk1
- Entrez Gene ID
- 25238
- Gene Name
- adrenergic, beta, receptor kinase 1
- Gene Aliases
- BARK1, GRK-2, Grk2
- Chromosome Location
- Chr. 1: 219536220 - 219544329 on Build Rnor_6.0
- UniGene ID
- Rn.13010
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012776.1 |
NP_036908.1
|
4 |
556 |
|
GenBank mRNA |
M87854.1 |
- |
4 |
556 |
|
View Full Product Details