Gene Transcripts

Gene Symbol
Ppm1a
Entrez Gene ID
24666
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1A
Gene Aliases
Pp2c1
Chromosome Location
Chr. 6: 94433661 - 94490408 on Build Rnor_6.0
UniGene ID
Rn.37403
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017038.1 NP_058734.1 2 405
XM_006240193.2 XP_006240255.1 2 3708
XM_006240194.2 XP_006240256.1 2 662
XM_006240195.2 XP_006240257.1 2 426
GenBank mRNA J04503.1 - 2 405

View Full Product Details