Gene Transcripts

Gene Symbol
KIN
Entrez Gene ID
22944
Gene Name
Kin17 DNA and RNA binding protein
Gene Aliases
BTCD, KIN17, Rts2
Chromosome Location
Chr. 10: 7750962 - 7788027 on Build GRCh38
UniGene ID
Hs.397918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012311.3 NP_036443.1 10, 11 1008
NR_045609.1 10, 11 1008
XM_006717434.3 XP_006717497.1 10, 11 1003
XM_006717435.3 XP_006717498.1 9, 10 876
XM_011519427.1 XP_011517729.1 10, 11 1003
XM_017015946.1 XP_016871435.1 9, 10 879
GenBank mRNA AJ005273.1 - 10, 11 979
AK296637.1 - 9, 10 876
AK301789.1 - 10, 11 1008
BC017309.2 - 10, 11 925 5089578

View Full Product Details