Gene Transcripts
- Gene Symbol
- Ascl2
- Entrez Gene ID
- 24209
- Gene Name
- achaete-scute family bHLH transcription factor 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 216154002 - 216156409 on Build Rnor_6.0
- UniGene ID
- Rn.10486
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031503.2 |
NP_113691.1
|
2 |
655 |
|
GenBank mRNA |
X53724.1 |
- |
2 |
599 |
|
View Full Product Details