Gene Transcripts

Gene Symbol
Slc45a1
Entrez Gene ID
246258
Gene Name
solute carrier family 45, member 1
Gene Aliases
Dnb5, Past-A, Pasta
Chromosome Location
Chr. 5: 167672134 - 167696331 on Build Rnor_6.0
UniGene ID
Rn.203315
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144747.3 NP_653348.3 2 222
GenBank mRNA AB075229.1 - 1 131
BC093599.1 - 2 222 7315729

View Full Product Details