Gene Transcripts

Gene Symbol
ZNF214
Entrez Gene ID
7761
Gene Name
zinc finger protein 214
Gene Aliases
BAZ-1, BAZ1
Chromosome Location
Chr. 11: 6997085 - 7020367 on Build GRCh38
UniGene ID
Hs.733636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013249.2 NP_037381.2 2 401
XM_005253128.3 XP_005253185.1 3 476
XM_005253129.3 XP_005253186.1 2 413
XM_006718308.3 XP_006718371.1 2 409
XM_006718309.3 XP_006718372.1 2 409
XM_011520355.2 XP_011518657.1 3 472
XM_011520357.2 XP_011518659.1 2 167
XM_017018269.1 XP_016873758.1 2 181
GenBank mRNA AF056617.1 - 2 356
AK313462.1 - 2 373
BC136772.1 - 2 281 9020762
BC144405.1 - 3 340 9052926
DB262997.1 - 2 401

View Full Product Details