Gene Transcripts

Gene Symbol
Ccr7
Entrez Gene ID
287673
Gene Name
chemokine (C-C motif) receptor 7
Gene Aliases
Chromosome Location
Chr. 10: 87057335 - 87067444 on Build Rnor_6.0
UniGene ID
Rn.229736
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199489.4 NP_955783.1 3 614
GenBank mRNA AY379972.1 - 3 617
BC089762.1 - 601 7381637

View Full Product Details