Gene Transcripts

Gene Symbol
NPBWR2
Entrez Gene ID
2832
Gene Name
neuropeptides B/W receptor 2
Gene Aliases
GPR8
Chromosome Location
Chr. 20: 64105830 - 64106831 on Build GRCh38
UniGene ID
Hs.248118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005286.2 NP_005277.2 1 380
GenBank mRNA BC067481.1 - 1 380
BC067482.1 - 1 380 6971826
BC067483.1 - 1 380 6971827

View Full Product Details