Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013325.4 | NP_037457.3 | 3 | 275 | |
NM_178326.2 | NP_847896.1 | 3 | 275 | ||
XM_005246992.3 | XP_005247049.2 | 3 | 261 | ||
XM_005246993.2 | XP_005247050.2 | 3 | 261 | ||
XM_005246995.3 | XP_005247052.1 | 3 | 249 | ||
XM_005246996.2 | XP_005247053.1 | 3 | 194 | ||
XM_017003637.1 | XP_016859126.1 | 3 | 374 | ||
XM_017003638.1 | XP_016859127.1 | 3 | 373 | ||
XM_017003639.1 | XP_016859128.1 | 2 | 177 | ||
XM_017003640.1 | XP_016859129.1 | 3 | 183 | ||
GenBank mRNA | AB023160.2 | - | 3 | 182 | |
AB066215.1 | - | 2 | 172 | ||
AJ504652.1 | - | 2 | 178 | ||
AK027332.1 | - | 2 | 127 | ||
AK027763.1 | - | 3 | 208 | ||
AK125277.1 | - | 3 | 699 | ||
AK296818.1 | - | 3 | 194 | ||
AK298109.1 | - | 3 | 194 | ||
AK302963.1 | - | 4 | 525 | ||
AL080168.1 | - | 3 | 276 | ||
BC000719.2 | - | 3 | 179 | 3504267 | |
BX648007.1 | - | 3 | 193 |