Gene Transcripts

Gene Symbol
OR3A3
Entrez Gene ID
8392
Gene Name
olfactory receptor family 3 subfamily A member 3
Gene Aliases
OR17-137, OR17-16, OR17-201, OR3A6, OR3A7, OR3A8P
Chromosome Location
Chr. 17: 3420424 - 3421805 on Build GRCh38
UniGene ID
Hs.532689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012373.2 NP_036505.2 1 798
XM_011524028.2 XP_011522330.1 2 808
GenBank mRNA BC069415.1 - 1 798
BC108921.2 - 1 792 40023068

View Full Product Details