Gene Transcripts

Gene Symbol
RASD2
Entrez Gene ID
23551
Gene Name
RASD family member 2
Gene Aliases
MGC:4834, Rhes, TEM2
Chromosome Location
Chr. 22: 35540492 - 35554003 on Build GRCh38
UniGene ID
Hs.474711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014310.3 NP_055125.2 3 1335
XM_005261442.4 XP_005261499.1 3 1195
XM_011530040.2 XP_011528342.1 4 2166
XM_017028701.1 XP_016884190.1 2 3944
XM_017028702.1 XP_016884191.1 3 2253
GenBank mRNA AF279143.1 - 2 1166
AL831936.1 - 2 1044
BC013419.2 - 3 1337 3604196

View Full Product Details