Gene Transcripts

Gene Symbol
RBPJL
Entrez Gene ID
11317
Gene Name
recombination signal binding protein for immunoglobulin kappa J region like
Gene Aliases
RBPL, RBPSUHL, SUHL
Chromosome Location
Chr. 20: 45306836 - 45319677 on Build GRCh38
UniGene ID
Hs.248217
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281448.1 NP_001268377.1 11 1351
NM_014276.3 NP_055091.2 11 1351
XM_011528522.2 XP_011526824.1 11 1263
GenBank mRNA AB024964.1 - 11 1271
AB026048.1 - 11 1271

View Full Product Details