Gene Transcripts

Gene Symbol
CRH
Entrez Gene ID
1392
Gene Name
corticotropin releasing hormone
Gene Aliases
CRF, CRH1
Chromosome Location
Chr. 8: 66176377 - 66186685 on Build GRCh38
UniGene ID
Hs.75294
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000756.3 NP_000747.1 2 1151
XM_017013090.1 XP_016868579.1 6 1489
XM_017013091.1 XP_016868580.1 6 1552
XM_017013092.1 XP_016868581.1 6 1490
XM_017013093.1 XP_016868582.1 6 1629
GenBank mRNA AK075431.1 - 2 1097
BC011031.1 - 2 973 4280754

View Full Product Details