Gene Transcripts
- Gene Symbol
- CACYBP
- Entrez Gene ID
- 27101
- Gene Name
- calcyclin binding protein
- Gene Aliases
- GIG5, PNAS-107, S100A6BP, SIP
- Chromosome Location
- Chr. 1: 174999435 - 175012027 on Build GRCh38
- UniGene ID
- Hs.508524
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007214.1 |
NP_001007215.1
|
3, 4 |
580 |
|
NM_014412.2 |
NP_055227.1
|
3, 4 |
765 |
|
XM_017001046.1 |
XP_016856535.1
|
3, 4 |
712 |
|
GenBank mRNA |
AF057356.1 |
- |
3, 4 |
454 |
|
AF275803.1 |
- |
3, 4 |
399 |
|
AF314752.1 |
- |
3, 4 |
464 |
|
AK093425.1 |
- |
3, 4 |
552 |
|
AK313278.1 |
- |
3, 4 |
556 |
|
AL035305.1 |
- |
3, 4 |
457 |
|
AY423723.1 |
- |
3, 4 |
341 |
|
BC005975.1 |
- |
3, 4 |
566 |
4102572 |
BC022352.1 |
- |
3, 4 |
463 |
4281339 |
BC067823.1 |
- |
3, 4 |
447 |
5260416 |
BC078151.1 |
- |
3, 4 |
430 |
6065872 |
BQ432793.1 |
- |
3, 4 |
583 |
|
View Full Product Details