Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321291.1 | NP_001308220.1 | 4 | 575 | |
XM_005257542.2 | XP_005257599.1 | 4 | 599 | ||
XM_005257543.2 | XP_005257600.1 | 4 | 599 | ||
XM_011525065.1 | XP_011523367.1 | 4 | 610 | ||
XM_011525066.2 | XP_011523368.1 | 4 | 472 | ||
XM_011525067.2 | XP_011523369.1 | 4 | 472 | ||
XM_011525068.1 | XP_011523370.1 | 4 | 513 | ||
XM_011525069.2 | XP_011523371.1 | 4 | 472 | ||
XM_011525070.1 | XP_011523372.1 | 4 | 472 | ||
XM_011525071.2 | XP_011523373.1 | 4 | 472 | ||
XM_017024900.1 | XP_016880389.1 | 4 | 599 | ||
XM_017024901.1 | XP_016880390.1 | 2 | 134 | ||
GenBank mRNA | AB040886.1 | - | 4 | 492 | |
AK001671.1 | - | 4 | 610 | ||
AK022913.1 | - | 4 | 517 | ||
AK225155.1 | - | 4 | 517 | ||
AK225426.1 | - | 4 | 474 | ||
AY169386.1 | - | 2 | 274 | ||
BC016487.1 | - | 4 | 576 | ||
BC027992.1 | - | 4 | 513 | ||
BC038983.1 | - | 4 | 584 | ||
BC071582.1 | - | 4 | 575 | 30345255 |