Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006837.2 | NP_006828.2 | 3 | 829 | |
GenBank mRNA | AB209134.1 | - | 2 | 2539 | |
AK056967.1 | - | 5 | 605 | ||
AK128519.1 | - | 2 | 2821 | ||
AK310434.1 | - | 3 | 631 | ||
AK311071.1 | - | 3 | 633 | ||
BC001187.1 | - | 3 | 633 | 3354130 | |
BC001859.2 | - | 3 | 611 | 3530890 | |
BC007272.1 | - | 3 | 607 | 3161025 | |
BX648542.1 | - | 3 | 3166 | ||
CR541678.1 | - | 3 | 498 | ||
U65928.1 | - | 3 | 625 | ||
U70734.1 | - | 3 | 618 |