Gene Transcripts

Gene Symbol
FFAR1
Entrez Gene ID
2864
Gene Name
free fatty acid receptor 1
Gene Aliases
FFA1R, GPCR40, GPR40
Chromosome Location
Chr. 19: 35351542 - 35352464 on Build GRCh38
UniGene ID
Hs.248127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005303.2 NP_005294.1 1 608
GenBank mRNA BC095536.1 - 1 595
BC120943.1 - 1 595
BC120944.1 - 1 598 40115111
EF611308.1 - 1 598

View Full Product Details