Gene Transcripts

Gene Symbol
RBP2
Entrez Gene ID
5948
Gene Name
retinol binding protein 2
Gene Aliases
CRABP-II, CRBP2, CRBPII, RBPC2
Chromosome Location
Chr. 3: 139452884 - 139476510 on Build GRCh38
UniGene ID
Hs.655516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004164.2 NP_004155.2 2, 3 299
GenBank mRNA AK291978.1 - 2, 3 305
BC069296.1 - 2, 3 248 7262683
BC069361.1 - 2, 3 248 7262695
BC069396.1 - 2, 3 248 7262707
BC069424.1 - 2, 3 248 7262719
BC069447.1 - 2, 3 248 7262731
BC069469.1 - 2, 3 247
BC069513.1 - 2, 3 248 7262755
BC069522.1 - 2, 3 248 7262672
BX282005.1 - 4, 5 426
U13831.1 - 2, 3 248

View Full Product Details