Gene Transcripts

Gene Symbol
SUPT16H
Entrez Gene ID
11198
Gene Name
SPT16 homolog, facilitates chromatin remodeling subunit
Gene Aliases
CDC68, FACTP140, SPT16, SPT16/CDC68
Chromosome Location
Chr. 14: 21351472 - 21384266 on Build GRCh38
UniGene ID
Hs.213724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007192.3 NP_009123.1 15 2114
XM_011536381.2 XP_011534683.1 16 2116
GenBank mRNA AK001719.1 - 15 1850
BC000565.2 - 15 1851
BC014046.2 - 15 1851
BC073849.1 - 15 1848

View Full Product Details