Gene Transcripts

Gene Symbol
MEX3B
Entrez Gene ID
84206
Gene Name
mex-3 RNA binding family member B
Gene Aliases
MEX-3B, RKHD3, RNF195
Chromosome Location
Chr. 15: 82041778 - 82046143 on Build GRCh38
UniGene ID
Hs.104744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032246.4 NP_115622.2 2 3368
GenBank mRNA AB095929.1 - 1 4180
AK094145.1 - 1 2602
AK094633.1 - 1 2032
AK131424.1 - 2 3245

View Full Product Details