Gene Transcripts
- Gene Symbol
- MYH14
- Entrez Gene ID
- 79784
- Gene Name
- myosin, heavy chain 14, non-muscle
- Gene Aliases
- DFNA4, DFNA4A, FP17425, MHC16, MYH17, NMHC II-C, NMHC-II-C, PNMHH, myosin
- Chromosome Location
- Chr. 19: 50188186 - 50310544 on Build GRCh38
- UniGene ID
- Hs.467142
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077186.1 |
NP_001070654.1
|
18, 19 |
2308 |
|
NM_001145809.1 |
NP_001139281.1
|
19, 20 |
2407 |
|
NM_024729.3 |
NP_079005.3
|
17, 18 |
2284 |
|
XM_006723386.3 |
XP_006723449.1
|
18, 19 |
2369 |
|
XM_011527320.2 |
XP_011525622.1
|
18, 19 |
2386 |
|
XM_011527321.2 |
XP_011525623.1
|
17, 18 |
2362 |
|
XM_011527323.2 |
XP_011525625.1
|
18, 19 |
2376 |
|
GenBank mRNA |
AB111886.1 |
- |
7, 8 |
966 |
|
AB290169.1 |
- |
18, 19 |
2377 |
|
AK125082.1 |
- |
18, 19 |
2387 |
|
AK307783.1 |
- |
18, 19 |
2290 |
|
AY165122.1 |
- |
17, 18 |
2284 |
|
BC018933.2 |
- |
16, 17 |
2036 |
|
FJ041910.1 |
- |
5, 6 |
570 |
|
View Full Product Details