Gene Transcripts

Gene Symbol
TNP2
Entrez Gene ID
7142
Gene Name
transition protein 2 (during histone to protamine replacement)
Gene Aliases
TP2
Chromosome Location
Chr. 16: 11267857 - 11269303 on Build GRCh38
UniGene ID
Hs.513349
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005425.4 NP_005416.1 1 418
GenBank mRNA BC096135.3 - 1 396
BC096136.3 - 1 396 40002550
BC096137.1 - 1 396 40002553
BX282422.1 - 1 418
X63758.1 - 1 377
X63759.1 - 1 645

View Full Product Details