Gene Transcripts

Gene Symbol
GAS1
Entrez Gene ID
2619
Gene Name
growth arrest specific 1
Gene Aliases
Chromosome Location
Chr. 9: 86944362 - 86947189 on Build GRCh38
UniGene ID
Hs.65029
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002048.2 NP_002039.2 1 1204
GenBank mRNA BC074809.2 - 1 836
BC074908.2 - 1 836 30915255
BC132682.1 - 1 879 40146704
BC136586.1 - 1 879 9020576
L13698.1 - 1 1204

View Full Product Details