Gene Transcripts

Gene Symbol
OR51V1
Entrez Gene ID
283111
Gene Name
olfactory receptor family 51 subfamily V member 1
Gene Aliases
OR11-36, OR51A12
Chromosome Location
Chr. 11: 5199735 - 5200700 on Build GRCh38
UniGene ID
Hs.553670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004760.2 NP_001004760.2 1 52

View Full Product Details