Gene Transcripts
- Gene Symbol
- DPH3
- Entrez Gene ID
- 285381
- Gene Name
- diphthamide biosynthesis 3
- Gene Aliases
- DELGIP, DELGIP1, DESR1, DPH3A, KTI11, ZCSL2
- Chromosome Location
- Chr. 3: 16257061 - 16264989 on Build GRCh38
- UniGene ID
- Hs.388087
- Species
- Homo sapiens
- Gene Symbol
- DPH3P1
- Entrez Gene ID
- 100132911
- Gene Name
- diphthamide biosynthesis 3 pseudogene 1
- Gene Aliases
- C20orf143, DPH3B, ZCSL1
- Chromosome Location
- Chr. 20: 62844566 - 62846191 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080750.4 |
NP_542788.1
|
1 |
1515 |
|
View Full Product Details