Gene Transcripts

Gene Symbol
AAMDC
Entrez Gene ID
28971
Gene Name
adipogenesis associated Mth938 domain containing
Gene Aliases
C11orf67, CK067, PTD015
Chromosome Location
Chr. 11: 77821114 - 77901281 on Build GRCh38
UniGene ID
Hs.745040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316957.1 NP_001303886.1 3 255
NM_001316958.1 NP_001303887.1 3 255
NM_001316960.1 NP_001303889.1 3 331
NM_001316961.1 NP_001303890.1 3 251
NM_001316962.1 NP_001303891.1 3 255
NM_024684.3 NP_078960.1 2 213
NR_133637.1 2 213
XM_011544968.2 XP_011543270.1 2 213
XM_011544969.2 XP_011543271.1 2 206
XM_017017620.1 XP_016873109.1 2 235
GenBank mRNA AF092136.1 - 3 214
AK024688.1 - 2 172
BC002752.2 - 2 165 3628061
BC016854.2 - 2 142 3856546
BI791786.1 - 3 264
BM930331.1 - 3 200
BP270361.1 - 2 213
BX646013.1 - 2 197
CN362204.1 - 3 226
CR457332.1 - 1 67

View Full Product Details