Gene Transcripts

Gene Symbol
NAV2
Entrez Gene ID
89797
Gene Name
neuron navigator 2
Gene Aliases
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location
Chr. 11: 19345200 - 20121601 on Build GRCh38
UniGene ID
Hs.502116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111018.1 NP_001104488.1 14 3775
NM_001111019.2 NP_001104489.1 5 1253
NM_001244963.1 NP_001231892.1 15 4168
NM_145117.4 NP_660093.2 14 4099
NM_182964.5 NP_892009.3 14 4099
XM_005253214.4 XP_005253271.1 4 5685
XM_006718364.3 XP_006718427.1 14 4421
XM_006718365.3 XP_006718428.1 15 4490
XM_006718366.3 XP_006718429.1 15 3654
XM_006718367.3 XP_006718430.1 4 5688
XM_006718368.3 XP_006718431.1 4 5685
XM_006718369.2 XP_006718432.1 4 1183
XM_011520444.2 XP_011518746.1 15 4490
XM_011520445.2 XP_011518747.1 15 4490
XM_011520446.2 XP_011518748.1 15 4490
XM_011520447.2 XP_011518749.1 15 4490
XM_011520448.2 XP_011518750.1 15 4361
XM_011520449.2 XP_011518751.1 15 4490
XM_011520450.2 XP_011518752.1 15 4490
XM_011520451.2 XP_011518753.1 14 3868
XM_011520452.2 XP_011518754.1 15 4238
XM_011520453.2 XP_011518755.1 4 1118
XM_011520454.2 XP_011518756.1 4 1118
XM_017018520.1 XP_016874009.1 16 4282
XM_017018521.1 XP_016874010.1 15 4490
XM_017018522.1 XP_016874011.1 15 4238
XM_017018523.1 XP_016874012.1 15 5167
XM_017018524.1 XP_016874013.1 15 3990
XM_017018525.1 XP_016874014.1 9 2833
XM_017018526.1 XP_016874015.1 5 3469
GenBank mRNA AB037840.3 - 14 4100
AB063115.1 - 14 3775
AB063116.1 - 14 4068
AF466143.1 - 14 3814
AF466144.1 - 14 3814
AJ488102.1 - 15 3906
AK074287.1 - 5 1253
AK298346.1 - 4 1265
AK307574.1 - 4 1081
BC150290.1 - 14 4100 8860130

View Full Product Details