Gene Transcripts
- Gene Symbol
- NKIRAS2
- Entrez Gene ID
- 28511
- Gene Name
- NFKB inhibitor interacting Ras like 2
- Gene Aliases
- KBRAS2, kappaB-Ras2
- Chromosome Location
- Chr. 17: 42017576 - 42025641 on Build GRCh38
- UniGene ID
- Hs.632252
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001349.2 |
NP_001001349.1
|
2, 3 |
157 |
|
NM_001144927.1 |
NP_001138399.1
|
2, 3 |
261 |
|
NM_001144928.1 |
NP_001138400.1
|
2, 3 |
242 |
|
NM_001144929.1 |
NP_001138401.1
|
2, 3 |
242 |
|
NM_017595.5 |
NP_060065.2
|
2, 3 |
242 |
|
XM_005257251.1 |
XP_005257308.1
|
2, 3 |
269 |
|
GenBank mRNA |
AF229840.1 |
- |
1, 2 |
92 |
|
AK027265.1 |
- |
2, 3 |
233 |
|
AK054571.1 |
- |
2, 3 |
269 |
|
AK297975.1 |
- |
2, 3 |
210 |
|
BC007316.2 |
- |
2, 3 |
194 |
|
BC007450.2 |
- |
2, 3 |
150 |
3028708 |
BC063498.1 |
- |
2, 3 |
713 |
5248950 |
BG717103.1 |
- |
2, 3 |
265 |
|
DA509824.1 |
- |
2, 3 |
211 |
|
DA607452.1 |
- |
2, 3 |
240 |
|
View Full Product Details