Gene Transcripts

Gene Symbol
RBMS3
Entrez Gene ID
27303
Gene Name
RNA binding motif single stranded interacting protein 3
Gene Aliases
Chromosome Location
Chr. 3: 29280860 - 30010395 on Build GRCh38
UniGene ID
Hs.221436
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003792.2 NP_001003792.1 3 648
NM_001003793.2 NP_001003793.1 3 651
NM_001177711.1 NP_001171182.1 3 651
NM_001177712.1 NP_001171183.1 3 651
NM_014483.3 NP_055298.2 3 651
XM_005265060.2 XP_005265117.1 3 1100
XM_005265061.2 XP_005265118.1 3 1103
XM_005265062.1 XP_005265119.1 3 1103
XM_017006178.1 XP_016861667.1 3 1100
XM_017006179.1 XP_016861668.1 3 1100
XM_017006180.1 XP_016861669.1 3 1103
XM_017006181.1 XP_016861670.1 3 1100
XM_017006182.1 XP_016861671.1 4 1179
GenBank mRNA AF023259.1 - 3 428
AK292789.1 - 3 327
AY236871.1 - 3 511
BC117315.1 - 3 348 40125866
BC143521.1 - 3 348 9052030

View Full Product Details