Gene Transcripts

Gene Symbol
PGAP2
Entrez Gene ID
27315
Gene Name
post-GPI attachment to proteins 2
Gene Aliases
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location
Chr. 11: 3797724 - 3826371 on Build GRCh38
UniGene ID
Hs.133968
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145438.2 NP_001138910.1 7 915
NM_001256235.1 NP_001243164.1 8 868
NM_001256236.1 NP_001243165.1 8 1110
NM_001256237.1 NP_001243166.1 7 1006
NM_001256238.1 NP_001243167.1 7 914
NM_001256239.1 NP_001243168.1 6 845
NM_001256240.1 NP_001243169.1 6 857
NM_001283038.1 NP_001269967.1 7 927
NM_001283039.1 NP_001269968.1 7 880
NM_001283040.1 NP_001269969.1 5 499
NM_014489.3 NP_055304.1 7 1040
NR_027016.2 5 682
NR_027017.2 8 1214
NR_027018.2 8 879
NR_045923.1 6 752
NR_045925.1 6 662
NR_045926.1 6 674
NR_045927.1 6 831
NR_045929.1 6 798
NR_104270.1 6 740
NR_104271.1 8 1000
NR_104272.1 5 670
XM_006718181.3 XP_006718244.1 6 1518
XM_006718183.3 XP_006718246.1 8 935
XM_006718185.2 XP_006718248.1 7 827
XM_006718186.2 XP_006718249.1 8 987
XM_006718190.3 XP_006718253.1 7 1143
XM_006718191.3 XP_006718254.1 7 1131
XM_006718192.1 XP_006718255.1 9 1061
XM_006718193.2 XP_006718256.1 8 868
XM_011519990.2 XP_011518292.2 8 1394
XM_011519991.2 XP_011518293.2 8 1385
XM_011519992.1 XP_011518294.1 9 1166
XM_011519993.2 XP_011518295.2 8 1492
XM_011519994.2 XP_011518296.2 8 1486
XM_011519995.2 XP_011518297.1 8 1317
XM_011519996.1 XP_011518298.1 9 1118
XM_011519997.1 XP_011518299.1 9 1459
XM_011519998.2 XP_011518300.1 9 1137
XM_011519999.1 XP_011518301.1 9 1096
XM_011520000.1 XP_011518302.1 8 1010
XM_011520001.1 XP_011518303.1 7 802
XM_011520002.1 XP_011518304.1 7 790
XM_011520003.2 XP_011518305.1 8 1326
XM_011520004.2 XP_011518306.1 8 1001
XM_011520005.2 XP_011518307.1 7 898
XM_011520006.2 XP_011518308.1 8 1282
XM_011520007.2 XP_011518309.1 9 1051
XM_017017556.1 XP_016873045.1 7 881
XM_017017557.1 XP_016873046.1 7 950
XM_017017558.1 XP_016873047.1 7 1124
XM_017017559.1 XP_016873048.1 8 987
XM_017017560.1 XP_016873049.1 8 975
GenBank mRNA AF159615.1 - 7 1042
AK292181.1 - 8 1004
AK295202.1 - 8 879
AK301991.1 - 8 1184
AK307359.1 - 6 669
AL049261.1 - 3 2069
AL096753.1 - 8 868
AL558360.3 - 6 583
AY373030.1 - 5 731
BC009930.2 - 7 802 3506614
BG468212.1 - 6 551
BI759334.1 - 6 738
BI858175.1 - 2 199
BX340489.2 - 6 782
BX356273.2 - 7 834
BX448155.2 - 7 786
CD675009.1 - 4 393
CD675113.1 - 6 633
CX872317.1 - 5 610
KU178608.1 - 5 719
KU178609.1 - 6 889
KU178610.1 - 6 816
KU178611.1 - 5 731

View Full Product Details