Gene Transcripts

Gene Symbol
AATK-AS1
Entrez Gene ID
388428
Gene Name
AATK antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 17: 81165507 - 81183164 on Build GRCh38
UniGene ID
Hs.591202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027255.1 5, 6 1371
GenBank mRNA AK126811.1 - 5, 6 1371

View Full Product Details