Gene Transcripts
- Gene Symbol
- FCRL6
- Entrez Gene ID
- 343413
- Gene Name
- Fc receptor like 6
- Gene Aliases
- FcRH6
- Chromosome Location
- Chr. 1: 159800506 - 159816257 on Build GRCh38
- UniGene ID
- Hs.196955
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001004310.2 |
NP_001004310.2
|
5, 6 |
935 |
|
NM_001284217.1 |
NP_001271146.1
|
6, 7 |
1001 |
|
XM_005245128.3 |
XP_005245185.1
|
6, 7 |
993 |
|
XM_005245129.3 |
XP_005245186.1
|
5, 6 |
960 |
|
XM_005245131.3 |
XP_005245188.1
|
6, 7 |
1011 |
|
XM_006711292.3 |
XP_006711355.1
|
6, 7 |
1011 |
|
XM_011509480.1 |
XP_011507782.1
|
6, 7 |
1000 |
|
XM_011509481.2 |
XP_011507783.1
|
6, 7 |
1011 |
|
XM_017001176.1 |
XP_016856665.1
|
6, 7 |
864 |
|
XM_017001177.1 |
XP_016856666.1
|
5, 6 |
797 |
|
GenBank mRNA |
AK131201.1 |
- |
6, 7 |
1000 |
|
AY212514.1 |
- |
5, 6 |
901 |
|
AY513661.1 |
- |
5, 6 |
893 |
|
AY654627.1 |
- |
4, 5 |
609 |
|
AY654628.1 |
- |
5, 6 |
894 |
|
View Full Product Details