Gene Transcripts

Gene Symbol
PLEKHG7
Entrez Gene ID
440107
Gene Name
pleckstrin homology and RhoGEF domain containing G7
Gene Aliases
Chromosome Location
Chr. 12: 92736489 - 92772092 on Build GRCh38
UniGene ID
Hs.591013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004330.2 NP_001004330.1 12 2760
GenBank mRNA AK128530.1 - 12 2717

View Full Product Details