Gene Transcripts

Gene Symbol
LRRC52
Entrez Gene ID
440699
Gene Name
leucine rich repeat containing 52
Gene Aliases
Chromosome Location
Chr. 1: 165544241 - 165563948 on Build GRCh38
UniGene ID
Hs.664975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005214.3 NP_001005214.2 2 1086
GenBank mRNA AK098677.1 - 2 1322

View Full Product Details