Gene Transcripts

Gene Symbol
LRRC52
Entrez Gene ID
440699
Gene Name
leucine rich repeat containing 52
Gene Aliases
Chromosome Location
Chr. 1: 165544241 - 165563948 on Build GRCh38
UniGene ID
Hs.664975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005214.3 NP_001005214.2 1 513
GenBank mRNA AK098677.1 - 1 749
BC132981.1 - 1 485 40147003
BC132983.1 - 1 485 40147005

View Full Product Details