Gene Transcripts

Gene Symbol
LHFPL4
Entrez Gene ID
375323
Gene Name
lipoma HMGIC fusion partner-like 4
Gene Aliases
Chromosome Location
Chr. 3: 9498361 - 9553802 on Build GRCh38
UniGene ID
Hs.56782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198560.2 NP_940962.1 4 1105
XM_017006359.1 XP_016861848.1 3 1643
GenBank mRNA AY278320.1 - 4 1105
BC113964.1 - 4 841

View Full Product Details