Gene Transcripts

Gene Symbol
OR5AS1
Entrez Gene ID
219447
Gene Name
olfactory receptor family 5 subfamily AS member 1
Gene Aliases
OR11-168
Chromosome Location
Chr. 11: 56030419 - 56031393 on Build GRCh38
UniGene ID
Hs.553625
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001921.1 NP_001001921.1 1 234

View Full Product Details