Gene Transcripts

Gene Symbol
OR51M1
Entrez Gene ID
390059
Gene Name
olfactory receptor family 51 subfamily M member 1
Gene Aliases
HOR5'Beta7, OR11-40
Chromosome Location
Chr. 11: 5389377 - 5390434 on Build GRCh38
UniGene ID
Hs.553731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004756.2 NP_001004756.2 1 238
GenBank mRNA BC136912.1 - 1 238
BC136913.1 - 1 238 9020904

View Full Product Details