Gene Transcripts
- Gene Symbol
- CMIP
- Entrez Gene ID
- 80790
- Gene Name
- c-Maf inducing protein
- Gene Aliases
- TCMIP
- Chromosome Location
- Chr. 16: 81445170 - 81711762 on Build GRCh38
- UniGene ID
- Hs.594095
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030629.2 |
NP_085132.1
|
18, 19 |
1930 |
|
NM_198390.2 |
NP_938204.2
|
18, 19 |
2156 |
|
XM_005256179.4 |
XP_005256236.4
|
18, 19 |
3111 |
|
XM_005256181.2 |
XP_005256238.1
|
17, 18 |
1887 |
|
XM_005256182.1 |
XP_005256239.1
|
16, 17 |
1644 |
|
XM_011523352.1 |
XP_011521654.1
|
17, 18 |
2068 |
|
XM_011523353.1 |
XP_011521655.1
|
16, 17 |
2024 |
|
XM_017023733.1 |
XP_016879222.1
|
16, 17 |
6501 |
|
GenBank mRNA |
AB051481.1 |
- |
18, 19 |
2037 |
|
AK096598.1 |
- |
18, 19 |
2117 |
|
AK304418.1 |
- |
17, 18 |
1892 |
|
AY172689.1 |
- |
18, 19 |
1930 |
|
BC038113.1 |
- |
16, 17 |
1853 |
4818904 |
View Full Product Details