Gene Transcripts

Gene Symbol
Smpd1
Entrez Gene ID
308909
Gene Name
sphingomyelin phosphodiesterase 1, acid lysosomal
Gene Aliases
Chromosome Location
Chr. 1: 170383682 - 170387525 on Build Rnor_6.0
UniGene ID
Rn.18277
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006997.1 NP_001006998.1 1 182
GenBank mRNA BC083780.1 - 1 182

View Full Product Details