Gene Transcripts

Gene Symbol
Wipi2
Entrez Gene ID
288498
Gene Name
WD repeat domain, phosphoinositide interacting 2
Gene Aliases
Chromosome Location
Chr. 12: 13969704 - 13998134 on Build Rnor_6.0
UniGene ID
Rn.163194
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007615.1 NP_001007616.1 4, 5 624
GenBank mRNA BC079184.1 - 4, 5 624

View Full Product Details