Gene Transcripts
- Gene Symbol
- TM9SF4
- Entrez Gene ID
- 9777
- Gene Name
- transmembrane 9 superfamily member 4
- Gene Aliases
- dJ836N17.2
- Chromosome Location
- Chr. 20: 32109506 - 32167258 on Build GRCh38
- UniGene ID
- Hs.654665
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014742.3 |
NP_055557.2
|
3, 4 |
459 |
|
XM_005260622.4 |
XP_005260679.1
|
3, 4 |
511 |
|
XM_017028154.1 |
XP_016883643.1
|
3, 4 |
240 |
|
XM_017028155.1 |
XP_016883644.1
|
3, 4 |
511 |
|
XM_017028159.1 |
XP_016883648.1
|
3, 4 |
292 |
|
XM_017028161.1 |
XP_016883650.1
|
3, 4 |
240 |
|
GenBank mRNA |
AK126534.1 |
- |
3, 4 |
251 |
|
AK296500.1 |
- |
3, 4 |
239 |
|
AK316293.1 |
- |
2, 3 |
163 |
|
BC021107.1 |
- |
3, 4 |
235 |
4896167 |
BC022850.2 |
- |
2, 3 |
217 |
|
D87444.1 |
- |
3, 4 |
513 |
|
DC381660.1 |
- |
3, 4 |
459 |
|
View Full Product Details