Gene Transcripts

Gene Symbol
Sertad1
Entrez Gene ID
361526
Gene Name
SERTA domain containing 1
Gene Aliases
Chromosome Location
Chr. 1: 84293102 - 84296370 on Build Rnor_6.0
UniGene ID
Rn.103853
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007735.1 NP_001007736.1 2 888
GenBank mRNA BC061808.1 - 2 888
FQ214111.1 - 2 902

View Full Product Details