Gene Transcripts

Gene Symbol
Eif1ad
Entrez Gene ID
293673
Gene Name
eukaryotic translation initiation factor 1A domain containing
Gene Aliases
RGD1304686
Chromosome Location
Chr. 1: 220746387 - 220751687 on Build Rnor_6.0
UniGene ID
Rn.4163
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008305.1 NP_001008306.1 6 1472
GenBank mRNA BC085818.1 - 6 1472

View Full Product Details