Gene Transcripts

Gene Symbol
Trappc3
Entrez Gene ID
362599
Gene Name
trafficking protein particle complex 3
Gene Aliases
Chromosome Location
Chr. 5: 144281720 - 144295306 on Build Rnor_6.0
UniGene ID
Rn.925
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008376.1 NP_001008377.1 1, 2 102
GenBank mRNA BC086377.1 - 1, 2 102
FQ214189.1 - 1, 2 158

View Full Product Details