Gene Transcripts

Gene Symbol
Ispd
Entrez Gene ID
493574
Gene Name
isoprenoid synthase domain containing
Gene Aliases
Nip
Chromosome Location
Chr. 6: 55880133 - 56159466 on Build Rnor_6.0
UniGene ID
Rn.119606
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008386.1 NP_001008387.1 7 1087
GenBank mRNA AY769991.1 - 6 772
AY769992.1 - 7 1087

View Full Product Details